Learn More
S2 Media 15x100mm Columbia Agar, 10/pack
Catalog No. 502995897
Encompass_Preferred
Columbia Agar is a non-selective and non-differential medium utilized for the isolation and cultivation of a wide variety of fastidious microorganisms, including Clostridium species. • Product meets CLSI, Harmonized USP/EP/JP, and USDA performance criteria.• Columbia Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Columbia Agar MLT (Microbial Limits Testing, USP)
This item is not returnable. View return policy
Catalog No. 50-299-5897 Supplier S2 Media Supplier No. 5307
Columbia Agar is a non-selective and non-differential medium utilized for the isolation and cultivation of a wide variety of fastidious microorganisms, including Clostridium species. • Product meets CLSI, Harmonized USP/EP/JP, and USDA performance criteria.• Columbia Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Columbia Agar MLT (Microbial Limits Testing, USP)