Learn More
S2 Media 15x100mm Columbia Blood Agar Base with Gentamicin, 10/pack
Catalog No. 502995796
Encompass_Preferred
Columbia Agar Base supplemented with 100µg/mL Gentamicin (without blood) is a selective and non-differential media used for the cultivation of fastidious and non-fastidious microorganisms. The medium is supplemented with gentamicin to suppress gram-negative bacteria present in sample flora.• Product meets CLSI reference methods and performance criteria.• Columbia Blood Agar Base w/ Gen-100 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5796 Supplier S2 Media Supplier No. 5051
Columbia Agar Base supplemented with 100µg/mL Gentamicin (without blood) is a selective and non-differential media used for the cultivation of fastidious and non-fastidious microorganisms. The medium is supplemented with gentamicin to suppress gram-negative bacteria present in sample flora.• Product meets CLSI reference methods and performance criteria.• Columbia Blood Agar Base w/ Gen-100 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.