Learn More
S2 Media 15x100mm Columbia Agar with 5% Sheep Blood (Columbia Blood Agar, CBA), 10/pack
Catalog No. 502995936
Encompass_Preferred
Columbia Agar with 5% Sheep Blood (defibrinated), also known as Columbia Blood Agar, is a non-selective, differential medium utilized for the isolation and cultivation of a wide variety of fastidious microorganisms. The medium is suitable for supporting the growth and determining hemolytic reactions of a variety of microorganisms.• Product meets CLSI performance criteria.• Columbia Agar with 5% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5936 Supplier S2 Media Supplier No. 5011
Columbia Agar with 5% Sheep Blood (defibrinated), also known as Columbia Blood Agar, is a non-selective, differential medium utilized for the isolation and cultivation of a wide variety of fastidious microorganisms. The medium is suitable for supporting the growth and determining hemolytic reactions of a variety of microorganisms.• Product meets CLSI performance criteria.• Columbia Agar with 5% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.