Learn More
S2 Media 15x100mm Columbia CNA Agar, 10/pack
Catalog No. 502995791
Encompass_Preferred
Columbia CNA Agar, supplemented with 5% defibrinated sheep blood, is a selective and differential medium utilized for the isolation and differentiation of Gram-positive microorganisms. • Product meets CLSI performance criteria.• Columbia CNA Agar with 5% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5791 Supplier S2 Media Supplier No. 5013
Columbia CNA Agar, supplemented with 5% defibrinated sheep blood, is a selective and differential medium utilized for the isolation and differentiation of Gram-positive microorganisms. • Product meets CLSI performance criteria.• Columbia CNA Agar with 5% Sheep Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.