Learn More
S2 Media 15x100mm Columbia CNA with TFG6, 10/pack
Catalog No. 502995848
Encompass_Preferred
Columbia CNA Agar, supplemented with 5% defibrinated sheep blood, Tween 80,Fosfomycin tromethamine, and D-glucose-6-phosphate (CCNA w/TFG6), is a selective and differential medium utilized for the isolation and differentiation of Corynebacterium kroppenstedtii. • Product meets CLSI performance criteria.• For in vitro diagnostic (IVD) use.• Columbia CNA with TFG6 should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5848 Supplier S2 Media Supplier No. 5370
Columbia CNA Agar, supplemented with 5% defibrinated sheep blood, Tween 80,Fosfomycin tromethamine, and D-glucose-6-phosphate (CCNA w/TFG6), is a selective and differential medium utilized for the isolation and differentiation of Corynebacterium kroppenstedtii. • Product meets CLSI performance criteria.• For in vitro diagnostic (IVD) use.• Columbia CNA with TFG6 should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.