Learn More
S2 Media 15x100mm Corn Meal Agar with Tween 80, Deep Fill, 10/pack
Catalog No. 502995775
Encompass_Preferred
Corn Meal Agar is a non-selective and non-differential culture medium used for the cultivation of fungi, particularly the production of chlamydospores by Candida albicans. Corn meal infusion provides the nutrients required for fungal growth and agar is the solidifying agent. Tween 80 enhances chlamydospore formation. • Product meets ASM, CDC and CLSI reference methods and performance criteria.• Corn Meal Agar with Tween 80 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5775 Supplier S2 Media Supplier No. 5339
Corn Meal Agar is a non-selective and non-differential culture medium used for the cultivation of fungi, particularly the production of chlamydospores by Candida albicans. Corn meal infusion provides the nutrients required for fungal growth and agar is the solidifying agent. Tween 80 enhances chlamydospore formation. • Product meets ASM, CDC and CLSI reference methods and performance criteria.• Corn Meal Agar with Tween 80 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.