Learn More
S2 Media 15x100mm Czapek Yeast Extract Agar (CYA), Deep Fill, 10/pack
Catalog No. 502995839
Encompass_Preferred
Czapek Yeast Extract Agar (CYA) is a selective and non-differential medium used for the isolation, cultivation, and morphological identification of fungi, particularly Aspergillus and Penicillium species. It supports the growth of filamentous fungi while inhibiting many bacteria due to its defined composition and low pH. CYA allows observation of colony morphology, pigmentation, and sporulation patterns important for fungal identification.• CYA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5839 Supplier S2 Media Supplier No. 5444
Czapek Yeast Extract Agar (CYA) is a selective and non-differential medium used for the isolation, cultivation, and morphological identification of fungi, particularly Aspergillus and Penicillium species. It supports the growth of filamentous fungi while inhibiting many bacteria due to its defined composition and low pH. CYA allows observation of colony morphology, pigmentation, and sporulation patterns important for fungal identification.• CYA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.