Learn More
S2 Media 15x100mm Inhibitory Mold Agar (IMA) with Gentamicin (and Chloramphenicol), Deep Fill, 10/pack
Catalog No. 502995879
Encompass_Preferred
Inhibitory Mold Agar (IMA) with Gentamicin (and Chloramphenicol) is a selective and non-differential medium used for the isolation and cultivation of Histoplasma capsulatum, dermatophytes, and other pathogenic fungi. IMA, developed by Ulrich, is a general-purpose medium for cultivating fungi and is used for the selective isolation of pathogenic yeasts and molds (e.g., Cryptococcus neoformans, Histoplasma capsulatum, Trichophyton spp.) from clinical samples with mixed microbial flora.• Product meets CLSI reference methods and performance criteria.• IMA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5879 Supplier S2 Media Supplier No. 5429
Inhibitory Mold Agar (IMA) with Gentamicin (and Chloramphenicol) is a selective and non-differential medium used for the isolation and cultivation of Histoplasma capsulatum, dermatophytes, and other pathogenic fungi. IMA, developed by Ulrich, is a general-purpose medium for cultivating fungi and is used for the selective isolation of pathogenic yeasts and molds (e.g., Cryptococcus neoformans, Histoplasma capsulatum, Trichophyton spp.) from clinical samples with mixed microbial flora.• Product meets CLSI reference methods and performance criteria.• IMA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.