Learn More
S2 Media 15x100mm K Agar, 10/pack
Catalog No. 502995911
Encompass_Preferred
K Agar is a selective and non-differential medium used for the isolation and cultivation of Alicyclobacillus species and acidophilic microorganisms in fruit juices. When incubated at 45ºC, K Agar predominantly supports the growth of A. acidoterrestris and limits the growth of other species of the same genus. • Product meets IFU reference methods and performance criteria.• K Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5911 Supplier S2 Media Supplier No. 5045
K Agar is a selective and non-differential medium used for the isolation and cultivation of Alicyclobacillus species and acidophilic microorganisms in fruit juices. When incubated at 45ºC, K Agar predominantly supports the growth of A. acidoterrestris and limits the growth of other species of the same genus. • Product meets IFU reference methods and performance criteria.• K Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.