Learn More
S2 Media 15x100mm LB Agar, Lennox, 10/pack
Catalog No. 502995790
Encompass_Preferred
LB Agar, Lennox is a non-selective and non-differential medium recommended for the cultivation and maintenance of recombinant strains of Escherichia coli. LB Agar, Lennox is a nutritionally rich media developed by Lennox for molecular genetics studies. This formulation contains 5.0g/L of sodium chloride, compared to 10.0g/L in LB Agar, Miller, and 0.5g/mL in Luria-Bertani Broth. • LB Agar, Lennox should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5790 Supplier S2 Media Supplier No. 5009
LB Agar, Lennox is a non-selective and non-differential medium recommended for the cultivation and maintenance of recombinant strains of Escherichia coli. LB Agar, Lennox is a nutritionally rich media developed by Lennox for molecular genetics studies. This formulation contains 5.0g/L of sodium chloride, compared to 10.0g/L in LB Agar, Miller, and 0.5g/mL in Luria-Bertani Broth. • LB Agar, Lennox should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.