Learn More
S2 Media 15x100mm LB Agar, Miller, with Chloramphenicol-30, 10/pack
Catalog No. 502995948
Encompass_Preferred
LB Agar, Miller, with Chloramphenicol-30 is a selective and non-differential medium recommended for the propagation and maintenance of recombinant strains of Escherichia coli. LB Agar, Miller is a nutritionally rich media developed by Miller for molecular genetics studies. This formulation contains 10.0g/L of sodium chloride, compared to 5.0g/L in LB Agar, Lennox. • LB Agar, Lennox should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product contains 30µg/mL Chloramphenicol
Catalog No. 50-299-5948 Supplier S2 Media Supplier No. 5356
LB Agar, Miller, with Chloramphenicol-30 is a selective and non-differential medium recommended for the propagation and maintenance of recombinant strains of Escherichia coli. LB Agar, Miller is a nutritionally rich media developed by Miller for molecular genetics studies. This formulation contains 10.0g/L of sodium chloride, compared to 5.0g/L in LB Agar, Lennox. • LB Agar, Lennox should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product contains 30µg/mL Chloramphenicol