Learn More
S2 Media 15x100mm Lin's Cupric Sulfate Medium (LCSM), 10/pack
Catalog No. 502995849
Encompass_Preferred
Lin's Cupric Sulfate Medium (LCSM) is a selective and non-differential medium used for the cultivation and isolation of wild yeast species. Cupric sulfate inhibits brewer's yeast (Saccharomyces cerevisiae).This formula was developed to isolate and identify beer spoilage yeasts commonly found in industrial fermentation processes. • Product meets ASBC reference methods and performance criteria.• LCSM should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.
Catalog No. 50-299-5849 Supplier S2 Media Supplier No. 5372
Lin's Cupric Sulfate Medium (LCSM) is a selective and non-differential medium used for the cultivation and isolation of wild yeast species. Cupric sulfate inhibits brewer's yeast (Saccharomyces cerevisiae).This formula was developed to isolate and identify beer spoilage yeasts commonly found in industrial fermentation processes. • Product meets ASBC reference methods and performance criteria.• LCSM should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.