Learn More
S2 Media 15x100mm m Endo Agar (LES), 10/pack
Catalog No. 502995845
Encompass_Preferred
m Endo Agar (LES) is a selective and differential medium used for the enumeration of coliforms in water by the membrane filtration method. The coliform group are used as indicators of fecal pollution in water, for assessing the effectiveness of water treatment and disinfection, and for monitoring water quality. Lauryl Tryptose Broth (LTB/LSB) can be used as a preliminary enrichment step, resulting in higher coliform counts. • Product meets APHA and EPA reference standards and performance criteria.• m-Endo Agar (LES) should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Lawrence Experimental Station (LES) Endo Agar.
Catalog No. 50-299-5845 Supplier S2 Media Supplier No. 5489
m Endo Agar (LES) is a selective and differential medium used for the enumeration of coliforms in water by the membrane filtration method. The coliform group are used as indicators of fecal pollution in water, for assessing the effectiveness of water treatment and disinfection, and for monitoring water quality. Lauryl Tryptose Broth (LTB/LSB) can be used as a preliminary enrichment step, resulting in higher coliform counts. • Product meets APHA and EPA reference standards and performance criteria.• m-Endo Agar (LES) should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Lawrence Experimental Station (LES) Endo Agar.