Learn More
S2 Media 15x100mm M9 Minimal Salts Agar without Glucose (M9 w/o Glu), 10/pack
Catalog No. 502995783
Encompass_Preferred
M9 Minimal Salts Agar without Glucose is a selective and non-differential growth medium used for the cultivation of Escherichia coli and other bacteria under controlled nutritional conditions. M9 Agar is used to study microbial physiology, metabolism, and genetics under nutrient-limited conditions. It enables the selection of mutants or plasmid-bearing bacteria through antibiotic resistance or nutrient complementation and is also used to test a bacterium's ability to synthesize essential compounds such as amino acids or nucleotides.• M9 Minimal Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5783 Supplier S2 Media Supplier No. 5433
M9 Minimal Salts Agar without Glucose is a selective and non-differential growth medium used for the cultivation of Escherichia coli and other bacteria under controlled nutritional conditions. M9 Agar is used to study microbial physiology, metabolism, and genetics under nutrient-limited conditions. It enables the selection of mutants or plasmid-bearing bacteria through antibiotic resistance or nutrient complementation and is also used to test a bacterium's ability to synthesize essential compounds such as amino acids or nucleotides.• M9 Minimal Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.