Learn More
S2 Media 15x100mm MacConkey (MAC) Agar, 10/pack
Catalog No. 502995827
Encompass_Preferred
MacConkey (MAC) Agar is a selective and differential medium used for the detection and isolation of Gram-Negative enteric bacilli. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies. Increased acid production results in the precipitation of bile salts, or a halo, around lactose fermenting bacteria. Non-lactose fermenting bacteria such as Salmonella spp. grow but remain colorless with no bile precipitate. • Product meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5827 Supplier S2 Media Supplier No. 5008
MacConkey (MAC) Agar is a selective and differential medium used for the detection and isolation of Gram-Negative enteric bacilli. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies. Increased acid production results in the precipitation of bile salts, or a halo, around lactose fermenting bacteria. Non-lactose fermenting bacteria such as Salmonella spp. grow but remain colorless with no bile precipitate. • Product meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.