Learn More
S2 Media 15x100mm MacConkey Agar with Levofloxacin-16, Cefotaxime-10, and Ampicillin-100, 10/pack
Catalog No. 502995858
Encompass_Preferred
MacConkey (MAC) Agar with Levofloxacin and Cefotaxime is a selective and differential medium used for the detection and isolation of Gram-negative enteric bacilli. Levofloxacin is an antibiotic used to inhibit the growth of Gram-positive organisms. Cefotaxime is used in screening for extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae. Ampicillin is a ß-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. • Product (base) meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Contains 16µg/mL Levofloxacin, 10µg/mL Cefotaxime, and 100µg/mL Ampicillin.
This item is not returnable. View return policy
Catalog No. 50-299-5858 Supplier S2 Media Supplier No. 5325
MacConkey (MAC) Agar with Levofloxacin and Cefotaxime is a selective and differential medium used for the detection and isolation of Gram-negative enteric bacilli. Levofloxacin is an antibiotic used to inhibit the growth of Gram-positive organisms. Cefotaxime is used in screening for extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae. Ampicillin is a ß-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. • Product (base) meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Contains 16µg/mL Levofloxacin, 10µg/mL Cefotaxime, and 100µg/mL Ampicillin.