Learn More
S2 Media 15x100mm MacConkey Agar with Levofloxacin-16 (MAC w/Lev-16), 10/pack
Catalog No. 502995820
Encompass_Preferred
MacConkey Agar with Levofloxacin-16 (MAC w/Lev-16) is a selective and differential medium used for the detection and isolation of Gram-negative enteric bacilli. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies. Increased acid production results in the precipitation of bile salts, or a halo, around lactose fermenting bacteria. Levofloxacin is an antibiotic used to inhibit the growth of Gram-positive organisms. • Product (base) meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MAC w/Lev-16 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product contains 16µg/mL Levofloxacin.
This item is not returnable. View return policy
Catalog No. 50-299-5820 Supplier S2 Media Supplier No. 5267
MacConkey Agar with Levofloxacin-16 (MAC w/Lev-16) is a selective and differential medium used for the detection and isolation of Gram-negative enteric bacilli. Escherichia coli can ferment lactose to produce acid which results in a pH drop. This is indicated by neutral red resulting in pink colonies. Increased acid production results in the precipitation of bile salts, or a halo, around lactose fermenting bacteria. Levofloxacin is an antibiotic used to inhibit the growth of Gram-positive organisms. • Product (base) meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MAC w/Lev-16 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product contains 16µg/mL Levofloxacin.