Learn More
S2 Media 15x100mm Sorbitol MacConkey (sMAC) Agar, 10/pack
Catalog No. 502995905
Encompass_Preferred
Sorbitol MacConkey (sMAC) Agar, also known as MacConkey Agar with Sorbitol, is a selective and differential medium used for the isolation of sorbitol-negative Escherichia coli. March and Ratnam introduced sMAC in 1986 for the isolation of verotoxin-producing (VTEC) Escherichia coli O157:H7 from clinical specimens. VTEC does not ferment sorbitol (i.e. - sorbitol-negative), while non-VTEC Escherichia coli are sorbitol-positive. Further identification and/or epidemiological typing should be performed on suspect colonies.• Product meets BAM, CLSI, ISO, and SMWW reference methods and performance criteria.• sMAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5905 Supplier S2 Media Supplier No. 5083
Sorbitol MacConkey (sMAC) Agar, also known as MacConkey Agar with Sorbitol, is a selective and differential medium used for the isolation of sorbitol-negative Escherichia coli. March and Ratnam introduced sMAC in 1986 for the isolation of verotoxin-producing (VTEC) Escherichia coli O157:H7 from clinical specimens. VTEC does not ferment sorbitol (i.e. - sorbitol-negative), while non-VTEC Escherichia coli are sorbitol-positive. Further identification and/or epidemiological typing should be performed on suspect colonies.• Product meets BAM, CLSI, ISO, and SMWW reference methods and performance criteria.• sMAC should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.