Learn More
S2 Media 15x100mm Malt Extract Agar (MEA), also known as Malt Extract Agar Yeasts and Molds (MEAYM), Deep Fill, 10/pack
Catalog No. 502995875
Encompass_Preferred
Malt Extract Agar (MEA), also known as Malt Extract Agar Yeasts and Molds (MEAYM), is a selective and non-differential medium used for the isolation, cultivation, and enumeration of yeasts and molds. The acidic pH of MEA inhibits bacterial growth while allowing optimal growth of fungi.• Product meets BAM and CLSI reference methods and performance criteria.• MEA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5875 Supplier S2 Media Supplier No. 5074
Malt Extract Agar (MEA), also known as Malt Extract Agar Yeasts and Molds (MEAYM), is a selective and non-differential medium used for the isolation, cultivation, and enumeration of yeasts and molds. The acidic pH of MEA inhibits bacterial growth while allowing optimal growth of fungi.• Product meets BAM and CLSI reference methods and performance criteria.• MEA should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.