Learn More
S2 Media 15x100mm Luria Broth Agar, 10/pack
Catalog No. 502995939
Encompass_Preferred
Cellulose Agar, Modified is a selective and non-differential medium used for the isolation , cultivation, and enumeration of cellulose-decomposing microorganisms. • Product meets ASM and CLSI reference methods and performance criteria.• Modified Cellulose Agar should be used by trained professionals within a laboratory setting. • For in Virto diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5939 Supplier S2 Media Supplier No. 5421
Cellulose Agar, Modified is a selective and non-differential medium used for the isolation , cultivation, and enumeration of cellulose-decomposing microorganisms. • Product meets ASM and CLSI reference methods and performance criteria.• Modified Cellulose Agar should be used by trained professionals within a laboratory setting. • For in Virto diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.