Learn More
S2 Media 15x100mm Mueller Hinton II (MH II) Agar, 10/pack
Catalog No. 502995795
Encompass_Preferred
Mueller Hinton II (MH II) Agar is a non-selective and non-differential medium originally developed for the cultivation of pathogenic Neisseria; however, these organisms are now isolated on selective media. This media is manufactured to contain low levels of thymine and thymidine and controlled levels of calcium and magnesium.• Product meets BAM, CLSI, and WHO reference methods and performance criteria.• MH II Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5795 Supplier S2 Media Supplier No. 5043
Mueller Hinton II (MH II) Agar is a non-selective and non-differential medium originally developed for the cultivation of pathogenic Neisseria; however, these organisms are now isolated on selective media. This media is manufactured to contain low levels of thymine and thymidine and controlled levels of calcium and magnesium.• Product meets BAM, CLSI, and WHO reference methods and performance criteria.• MH II Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.