Learn More
S2 Media 15x100mm Nutrient Agar (NA), 10/pack
Catalog No. 502995869
Encompass_Preferred
Nutrient Agar (NA), is a non-selective and non-differential medium. It is recommended for acquiring microbial plate counts from dairy products, food, water, wastewater, and samples of sanitary importance.• Product meets AOAC, BAM, ISO, NF, SMWW, and USDA reference methods and performance criteria.• Nutrient Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5869 Supplier S2 Media Supplier No. 5006
Nutrient Agar (NA), is a non-selective and non-differential medium. It is recommended for acquiring microbial plate counts from dairy products, food, water, wastewater, and samples of sanitary importance.• Product meets AOAC, BAM, ISO, NF, SMWW, and USDA reference methods and performance criteria.• Nutrient Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.