Learn More
S2 Media 15x100mm Phenylethyl Alcohol (PEA) Agar, 10/pack
Catalog No. 502995969
Encompass_Preferred
Phenylethyl Alcohol (PEA) Agar is a selective and non-differential medium used for the isolation of gram-positive cocci, including Staphylococci and Streptococci. This medium is not suitable for hemolytic determinations due to the occurrence of atypical reactions. Phenylethyl alcohol is bacteriostatic for gram-negative bacteria.• Product meets CLSI and NF reference methods and performance criteria.• PEA Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5969 Supplier S2 Media Supplier No. 5036
Phenylethyl Alcohol (PEA) Agar is a selective and non-differential medium used for the isolation of gram-positive cocci, including Staphylococci and Streptococci. This medium is not suitable for hemolytic determinations due to the occurrence of atypical reactions. Phenylethyl alcohol is bacteriostatic for gram-negative bacteria.• Product meets CLSI and NF reference methods and performance criteria.• PEA Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.