Learn More
S2 Media 15x100mm Phenylethyl Alcohol Agar with 5% Sheep Blood (PEA w/ Blood), 10/pack
Catalog No. 502995832
Encompass_Preferred
Phenylethyl Alcohol Agar with 5% Sheep Blood (PEA w/ Blood) is a selective and non-differential medium used for the isolation of gram-positive cocci, including Staphylococci and Streptococci. This medium is not suitable for hemolytic determinations due to the occurrence of atypical reactions. Phenylethyl alcohol is bacteriostatic for gram-negative bacteria.• Product meets CLSI and NF reference methods and performance criteria.• PEA w/ Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5832 Supplier S2 Media Supplier No. 5037
Phenylethyl Alcohol Agar with 5% Sheep Blood (PEA w/ Blood) is a selective and non-differential medium used for the isolation of gram-positive cocci, including Staphylococci and Streptococci. This medium is not suitable for hemolytic determinations due to the occurrence of atypical reactions. Phenylethyl alcohol is bacteriostatic for gram-negative bacteria.• Product meets CLSI and NF reference methods and performance criteria.• PEA w/ Blood should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.