Learn More
S2 Media 15x100mm Standard Methods Agar (SMA) / Plate Count Agar (PCA) / Tryptone Glucose Yeast (TGY) Agar, 10/pack
Catalog No. 502995963
Encompass_Preferred
Standard Method Agar (SMA), also known as Plate Count Agar (PCA) or Tryptone Glucose Yeast (TGY) Agar, is a non-selective and non-differential medium. It is recommended for acquiring microbial plate counts from cosmetics, dairy products, food, water, wastewater, and samples of sanitary importance.• Product meets AOAC, APHA, BAM, EPA, Harmonized USP/EP/JP, ISO, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• SMA/PCA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5963 Supplier S2 Media Supplier No. 5005
Standard Method Agar (SMA), also known as Plate Count Agar (PCA) or Tryptone Glucose Yeast (TGY) Agar, is a non-selective and non-differential medium. It is recommended for acquiring microbial plate counts from cosmetics, dairy products, food, water, wastewater, and samples of sanitary importance.• Product meets AOAC, APHA, BAM, EPA, Harmonized USP/EP/JP, ISO, NF, SM-Dairy, SMWW, and USDA reference methods and performance criteria.• SMA/PCA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.