Learn More
S2 Media 15x100mm Potato Dextrose Agar with Tartaric Acid (PDA w/TA), Deep Fill, 10/pack
Catalog No. 502995943
Encompass_Preferred
Potato Dextrose Agar with Tartaric Acid (PDA w/TA) is a selective and non-differential medium used for the microbial examination of food and dairy products. • Product meets CLSI and Harmonized USP/EP/JP (base) reference methods and performance criteria.• PDA w/TA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5943 Supplier S2 Media Supplier No. 5054
Potato Dextrose Agar with Tartaric Acid (PDA w/TA) is a selective and non-differential medium used for the microbial examination of food and dairy products. • Product meets CLSI and Harmonized USP/EP/JP (base) reference methods and performance criteria.• PDA w/TA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.