Learn More
S2 Media 15x100mm Peptone Yeast Calcium (PYCa) Agar, 10/pack
Catalog No. 502995735
Encompass_Preferred
Peptone Yeast Calcium (PYCa) Agar is a general purpose medium utilized for most species of the bacterial genera Nocardia, Corynebacterium, Gordonia, Rhondococcus, Tsukamurella, and Mycobacterium for Phagehunting protocols. • Product meets Phagehunting Protocols (phagesdb.org) performance criteria.• PYCa should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5735 Supplier S2 Media Supplier No. 5139
Peptone Yeast Calcium (PYCa) Agar is a general purpose medium utilized for most species of the bacterial genera Nocardia, Corynebacterium, Gordonia, Rhondococcus, Tsukamurella, and Mycobacterium for Phagehunting protocols. • Product meets Phagehunting Protocols (phagesdb.org) performance criteria.• PYCa should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.