Learn More
S2 Media 15x100mm RPMI Agar with 2% MOPS and Glucose, Deep Fill, 10/pack
Catalog No. 502995872
Encompass_Preferred
RPMI Agar with 2% MOPS and Glucose is a non-selective, non-differential media utilized for Candida testing directly from colonies grown on non-selective media. • Product meets CLSI reference methods and performance criteria.• RPMI Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5872 Supplier S2 Media Supplier No. 5052
RPMI Agar with 2% MOPS and Glucose is a non-selective, non-differential media utilized for Candida testing directly from colonies grown on non-selective media. • Product meets CLSI reference methods and performance criteria.• RPMI Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.