Learn More
S2 Media 15x100mm Sabouraud Brain Heart Infusion (SaBHI) Agar with Chloramphenicol and Cycloheximide, Deep Fill, 10/pack
Catalog No. 502995921
Encompass_Preferred
S2Media™ Sabouraud Brain Heart Infusion (SaBHI) Agar with Chloramphenicol and Cycloheximide is a selective and non-differential medium used for the cultivation of dermatophytes and other fungi. Gorman developed SaBHI in 1967, by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. • Product meets ASM and CLSI reference methods and performance criteria.• SaBHI should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5921 Supplier S2 Media Supplier No. 5343
S2Media™ Sabouraud Brain Heart Infusion (SaBHI) Agar with Chloramphenicol and Cycloheximide is a selective and non-differential medium used for the cultivation of dermatophytes and other fungi. Gorman developed SaBHI in 1967, by combining Sabouraud Dextrose Agar and Brain Heart Infusion Agar. • Product meets ASM and CLSI reference methods and performance criteria.• SaBHI should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.