Learn More
S2 Media 15x100mm Sabouraud Dextrose Agar with Chloramphenicol 100 (SabDex w/ CHL-100), Deep Fill, 10/pack
Catalog No. 502995800
Encompass_Preferred
Sabouraud Dextrose Agar with Chloramphenicol-100 a selective and non-differential medium used for the isolation and cultivation of yeasts and molds from clinical and non-clinical specimens. The acidic pH and high dextrose content favor fungal growth, while chloramphenicol suppresses bacterial contaminants. Commonly used for cultivating dermatophytes, Candida, and other pathogenic and saprophytic fungi. Chloramphenicol is added to suppress bacterial contaminants that could otherwise overgrow and obscure fungal colonies.• Product (base) meets BAM, CLSI, Harmonized USP/EP/JP, ISO, NF, and SMWW reference methods and performance criteria.• SabDex Agar w/CHL-100 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5800 Supplier S2 Media Supplier No. 5434
Sabouraud Dextrose Agar with Chloramphenicol-100 a selective and non-differential medium used for the isolation and cultivation of yeasts and molds from clinical and non-clinical specimens. The acidic pH and high dextrose content favor fungal growth, while chloramphenicol suppresses bacterial contaminants. Commonly used for cultivating dermatophytes, Candida, and other pathogenic and saprophytic fungi. Chloramphenicol is added to suppress bacterial contaminants that could otherwise overgrow and obscure fungal colonies.• Product (base) meets BAM, CLSI, Harmonized USP/EP/JP, ISO, NF, and SMWW reference methods and performance criteria.• SabDex Agar w/CHL-100 should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.