Learn More
S2 Media 15x100mm Sabouraud Dextrose Agar (SDA, SabDex), Deep Fill, 10/pack
Catalog No. 502995828
Encompass_Preferred
Sabouraud Dextrose Agar (SDA) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. • Product meets BAM, CLSI, Harmonized USP/EP/JP, ISO, NF, and SMWW reference methods and performance criteria.• SabDex should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5828 Supplier S2 Media Supplier No. 5002
Sabouraud Dextrose Agar (SDA) is a selective and non-differential growth medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. • Product meets BAM, CLSI, Harmonized USP/EP/JP, ISO, NF, and SMWW reference methods and performance criteria.• SabDex should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.