Learn More
S2 Media 15x100mm Sabouraud Dextrose Agar, Emmons (Sabouraud Dextrose Agar, Modified), Deep Fill, 10/pack
Catalog No. 502995862
Encompass_Preferred
Sabouraud Dextrose Agar, Emmons, also known as Sabouraud Dextrose Agar, Modified, is a non-selective and non-differential general purpose medium used for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. The original formulation was modified by Emmons to bring the pH closer to neutral (pH range is 6.8-7.2) and the dextrose content was decrease from 40g/L to 20g/L. • Product meets CLSI and SMWW reference methods and performance criteria.• SabDex, Emmons should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5862 Supplier S2 Media Supplier No. 5341
Sabouraud Dextrose Agar, Emmons, also known as Sabouraud Dextrose Agar, Modified, is a non-selective and non-differential general purpose medium used for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. The original formulation was modified by Emmons to bring the pH closer to neutral (pH range is 6.8-7.2) and the dextrose content was decrease from 40g/L to 20g/L. • Product meets CLSI and SMWW reference methods and performance criteria.• SabDex, Emmons should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.