Learn More
S2 Media 15x100mm Sabouraud Dextrose Agar with Chloramphenicol-50 (SabDex w/ CHL-50), Deep Fill, 10/pack
Catalog No. 502995792
Encompass_Preferred
Sabouraud Dextrose Agar with Chloramphenicol-50 (SabDex w/ CHL-50) is a selective and non-differential medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. The medium is made selective by the addition of the antimicrobic, Chloramphenicol (50µg/mL), which inhibits Gram-Negative microorganisms. • Product (base) meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• Sabouraud Dextrose Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5792 Supplier S2 Media Supplier No. 5016
Sabouraud Dextrose Agar with Chloramphenicol-50 (SabDex w/ CHL-50) is a selective and non-differential medium utilized for the cultivation of pathogenic and non-pathogenic fungi, including dermatophytes. The medium is made selective by the addition of the antimicrobic, Chloramphenicol (50µg/mL), which inhibits Gram-Negative microorganisms. • Product (base) meets BAM, CLSI, ISO, NF, SMWW, and Harmonized USP/EP/JP reference methods and performance criteria.• Sabouraud Dextrose Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.