Learn More
S2 Media 15x100mm Salt Sabouraud Dulcitol (SSD) Agar, 10/pack
Catalog No. 502995763
Encompass_Preferred
Salt Sabouraud Dulcitol (SSD) Agar is a selective and non-differential medium used in qualitative screening procedures for the cultivation and selection of Candida auris, an emerging pathogen with multidrug resistance. Multiple methods are available and include use of a pre-enrichment step (utilizing SSD Broth) with subsequent subculture onto Salt Sabouraud Dulcitol Agar or a chromogenic medium such as Chameleon™ COLOREX™ Candida Plus. Direct plating methods are also available to enumerate and isolate colonies for additional identification testing.• Product meets CDC and CLSI reference methods and performance criteria.• SSDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5763 Supplier S2 Media Supplier No. 5194
Salt Sabouraud Dulcitol (SSD) Agar is a selective and non-differential medium used in qualitative screening procedures for the cultivation and selection of Candida auris, an emerging pathogen with multidrug resistance. Multiple methods are available and include use of a pre-enrichment step (utilizing SSD Broth) with subsequent subculture onto Salt Sabouraud Dulcitol Agar or a chromogenic medium such as Chameleon™ COLOREX™ Candida Plus. Direct plating methods are also available to enumerate and isolate colonies for additional identification testing.• Product meets CDC and CLSI reference methods and performance criteria.• SSDB should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.