Learn More
S2 Media 15x100mm Tryptic Soy Agar (TSA) / Soybean-Casein Digest (SCD) Agar, 10/pack
Catalog No. 502995937
Encompass_Preferred
Tryptic Soy Agar (TSA) is a non-selective and non-differential, general growth medium. It is recommended for the isolation and cultivation of fastidious and non-fastidious microorganisms, although it is not the medium of choice for anaerobes. TSA is also recommended for the preparation, maintenance, and storage of test strains. • Product complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5937 Supplier S2 Media Supplier No. 5000
Tryptic Soy Agar (TSA) is a non-selective and non-differential, general growth medium. It is recommended for the isolation and cultivation of fastidious and non-fastidious microorganisms, although it is not the medium of choice for anaerobes. TSA is also recommended for the preparation, maintenance, and storage of test strains. • Product complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.