Learn More
S2 Media 15x100mm Tryptic Soy Agar with 0.03% Congo Red (TSA w/CR), 10/pack
Catalog No. 502995823
Encompass_Preferred
Tryptic Soy Agar with 0.03% Congo Red (TSA w/CR) is a non-selective, differential medium used for the cultivation of a wide variety of non-fastidious microorganisms. Congo red is utilized to stain the cells of fungi and the outer membrane of Gram-negative bacteria. Additionally, studies have indicated virulent Shigella flexneri serotype 2a, grown in the presence of congo red at 37°C, showed increased levels of three proteins with MRS of 43K, 58K, and 63K proteins) in the Sarkosyl-soluble membrane fractions. The observed phenomenon is temperature dependent and is generally correlated with their virulence properties.• Product meets CLSI performance criteria.• TSA w/CR should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5823 Supplier S2 Media Supplier No. 5301
Tryptic Soy Agar with 0.03% Congo Red (TSA w/CR) is a non-selective, differential medium used for the cultivation of a wide variety of non-fastidious microorganisms. Congo red is utilized to stain the cells of fungi and the outer membrane of Gram-negative bacteria. Additionally, studies have indicated virulent Shigella flexneri serotype 2a, grown in the presence of congo red at 37°C, showed increased levels of three proteins with MRS of 43K, 58K, and 63K proteins) in the Sarkosyl-soluble membrane fractions. The observed phenomenon is temperature dependent and is generally correlated with their virulence properties.• Product meets CLSI performance criteria.• TSA w/CR should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.