Learn More
S2 Media 15x100mmTryptic Soy Agar with Lecithin and Tween 80 (TSA w/LT), Deep Fill, 10/pack
Catalog No. 502995931
Encompass_Preferred
Tryptic Soy Agar with Lecithin and Tween80 (TSA w/LT) is a non-selective and non-differential medium utilized for the detection and enumeration of microorganisms present on surfaces and in areas of sanitary importance. Prepared plates are utilized for environmental monitoring within biologically controlled environments. These plates are suitable for active and passive air sampling (settle/settling plates) as well as for personnel monitoring of fingertips (Finger Dabs). Lecithin is utilized to neutralize quaternary ammonium compounds. Polysorbate (Tween) 80 is used to neutralize substituted phenolic disinfectants. • Product meets USP-NF reference methods and performance criteria.• Deep fill (32mL).• TSA w/ LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Microbial Content Test Agar
This item is not returnable. View return policy
Catalog No. 50-299-5931 Supplier S2 Media Supplier No. 5186
Tryptic Soy Agar with Lecithin and Tween80 (TSA w/LT) is a non-selective and non-differential medium utilized for the detection and enumeration of microorganisms present on surfaces and in areas of sanitary importance. Prepared plates are utilized for environmental monitoring within biologically controlled environments. These plates are suitable for active and passive air sampling (settle/settling plates) as well as for personnel monitoring of fingertips (Finger Dabs). Lecithin is utilized to neutralize quaternary ammonium compounds. Polysorbate (Tween) 80 is used to neutralize substituted phenolic disinfectants. • Product meets USP-NF reference methods and performance criteria.• Deep fill (32mL).• TSA w/ LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Microbial Content Test Agar