Learn More
S2 Media 15x100mm Chameleon™ Tryptic Soy Agar with X-Gluc Sodium Salt (TSA w/X-Gluc)
Catalog No. 502995962
Encompass_Preferred
Chameleon™ Tryptic Soy Agar with X-Gluc Sodium Salt (TSA w/X-Gluc) differential chromogenic growth medium recommended for the detection of β-glucuronidase (GUD) activity, particularly for identifying Escherichia coli. Bacteria that produce β-glucuronidase will cleave X-Gluc, producing an insoluble blue compound. (These colonies appear blue to blue-green on the agar.)• Product base complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide) sodium salt
This item is not returnable. View return policy
Catalog No. 50-299-5962 Supplier S2 Media Supplier No. 5480
Chameleon™ Tryptic Soy Agar with X-Gluc Sodium Salt (TSA w/X-Gluc) differential chromogenic growth medium recommended for the detection of β-glucuronidase (GUD) activity, particularly for identifying Escherichia coli. Bacteria that produce β-glucuronidase will cleave X-Gluc, producing an insoluble blue compound. (These colonies appear blue to blue-green on the agar.)• Product base complies with AOAC, BAM, CLSI, ISO, Harmonized USP/EP/JP, SMWW, and USDA reference methods and performance criteria.• TSA should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide) sodium salt