Learn More
S2 Media 15x100mm Tryptic Soy Agar (TSA) with 5% Sheep Blood (Blood Agar), 10/pack
Catalog No. 502995807
Encompass_Preferred
Tryptic Soy Agar supplemented with 5% defibrinated sheep blood (TSA w/ 5% Sheep Blood, Blood Agar) is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. TSA w/ 5% Sheep Blood is not the medium of choice for anaerobes.• Product complies with BAM, CLSI, and USDA reference methods and performance criteria.• TSA with 5% Sheep Blood should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5807 Supplier S2 Media Supplier No. 5001
Tryptic Soy Agar supplemented with 5% defibrinated sheep blood (TSA w/ 5% Sheep Blood, Blood Agar) is a non-selective, differential growth medium utilized for the isolation and cultivation of non-fastidious and fastidious organisms. Hemolytic response can be utilized to differentiate microorganisms. TSA w/ 5% Sheep Blood is not the medium of choice for anaerobes.• Product complies with BAM, CLSI, and USDA reference methods and performance criteria.• TSA with 5% Sheep Blood should be used by trained professionals within a laboratory setting.• For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.