Learn More
S2 Media 15x100mm Universal Beer Agar (UBA), 10/pack
Catalog No. 502995851
Encompass_Preferred
Universal Beer Agar (UBA) is a selective and non-differential medium used for the cultivation of microorganisms of significance int he brewing industry. The presence of hops constituents and alcohol (i.e., beer) inhibit growth of many airborne microorganisms not adapted to the brewery environment.• Product meets ASBC reference methods and performance criteria.• UBA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.
Catalog No. 50-299-5851 Supplier S2 Media Supplier No. 5376
Universal Beer Agar (UBA) is a selective and non-differential medium used for the cultivation of microorganisms of significance int he brewing industry. The presence of hops constituents and alcohol (i.e., beer) inhibit growth of many airborne microorganisms not adapted to the brewery environment.• Product meets ASBC reference methods and performance criteria.• UBA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Not for use in diagnostic procedures.