Learn More
S2 Media 15x100mm Yeast Mold (YM) Agar / Yeast Malt Peptone Glucose (MYPG) Agar, Deep Fill, 10/pack
Catalog No. 502995901
Encompass_Preferred
Yeast Mold (YM) Agar is a selective and non-differential medium used for the cultivation of fungi and other aciduric microorganisms. This media is prepared according to the formula published by Wickerham and is not supplemented with antibiotics or fungistatic. This medium is also known as Yeast Malt Peptone Glucose (MYPG) Agar. • Product meets AOAC and NF reference methods and performance criteria.• YM Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5901 Supplier S2 Media Supplier No. 5029
Yeast Mold (YM) Agar is a selective and non-differential medium used for the cultivation of fungi and other aciduric microorganisms. This media is prepared according to the formula published by Wickerham and is not supplemented with antibiotics or fungistatic. This medium is also known as Yeast Malt Peptone Glucose (MYPG) Agar. • Product meets AOAC and NF reference methods and performance criteria.• YM Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.