Learn More
S2 Media 15x100mm Yeast Starch Glucose (YSG) Agar, 10/pack
Catalog No. 502995942
Encompass_Preferred
Yeast Starch Glucose (YSG) Agar is a selective and non-differential medium used for the isolation and cultivation of Alicyclobacillus species and acidophilic microorganisms in fruit juices and other acidic beverages or food. The low pH value of the media, in combination with the high incubation temperature, inhibits the growth of contaminating microbiota. • Product meets IFU reference methods and performance criteria.• YSG Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5942 Supplier S2 Media Supplier No. 5046
Yeast Starch Glucose (YSG) Agar is a selective and non-differential medium used for the isolation and cultivation of Alicyclobacillus species and acidophilic microorganisms in fruit juices and other acidic beverages or food. The low pH value of the media, in combination with the high incubation temperature, inhibits the growth of contaminating microbiota. • Product meets IFU reference methods and performance criteria.• YSG Agar should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.