Learn More
S2 Media 15x100mm Yersinia Selective Agar (CIN Agar), 10/pack
Supplier:  S2 Media 5402
Encompass_Preferred
S2Media™ Yersinia Selective Agar (Schiemann's CIN Agar) is a selective and differential medium used for isolation and enumeration of Yersinia species from food samples. Its formulation incorporates multiple selective agents, sodium deoxycholate, crystal violet, cefsulodin, irgasan, and novobiocin, to inhibit enteric flora. Yersinia spp. characteristically ferment mannitol, producing a concentrated acid reaction in the colony center that creates a distinctive red “bull’s-eye” morphology. In contrast, most other enteric bacteria, if they grow, form larger colonies with diffuse pink centers and opaque margins, allowing differentiation from Yersinia.• Product meets BAM, ISO, CLSI, SM-Dairy, USP-NF performance criteria.• Yersinia Selective Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5798