Learn More
S2 Media 15x150mm Haemophilus Test Medium (HTM), 5/pk
Catalog No. 502995908
Encompass_Preferred
S2Media™ Haemophilus Test Medium (HTM) Agar is a selective medium used for cultivation of fastidious microorganisms, including Haemophilus influenzae and Neisseria gonorrhoeae. It contains essential nutrients, such as supplements of yeast extract, starch, and defined growth factors (hemin and NAD), to support these fastidious organisms.• Product meets ASM and CLSI performance criteria.• HTM Agar should be used by trained professionals within a laboratory setting. • Product is not intended for IVD use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5908 Supplier S2 Media Supplier No. 5398
S2Media™ Haemophilus Test Medium (HTM) Agar is a selective medium used for cultivation of fastidious microorganisms, including Haemophilus influenzae and Neisseria gonorrhoeae. It contains essential nutrients, such as supplements of yeast extract, starch, and defined growth factors (hemin and NAD), to support these fastidious organisms.• Product meets ASM and CLSI performance criteria.• HTM Agar should be used by trained professionals within a laboratory setting. • Product is not intended for IVD use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.