Learn More
S2 Media 15x150mm Mueller-Hinton II Agar with 5% Sheep Blood, 5/pk
Catalog No. 502995780
Encompass_Preferred
S2Media™ Mueller-Hinton II (MH-II) Agar is a non-selective and non-differential medium. Originally developed by Mueller and Hinton as a transparent medium for isolating Neisseria species, MH-II uses starch in place of pea extract to protect against toxic substances. Bauer, Kirby, Sherris, and Tuck recommended the medium for antibiotic susceptibility testing. The Clinical and Laboratory Standards Institute (CLSI) endorses it because it supports consistent growth of most non-fastidious pathogens, minimizes inhibitors of key antibiotics, and ensures reliable batch-to-batch performance.• Product meets AOAC, ASM, BAM, CLSI, USDA, and WHO performance criteria.• Mueller-Hinton II Agar should be used by trained professionals within a laboratory setting. • Product is not intended for IVD use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5780 Supplier S2 Media Supplier No. 5399
S2Media™ Mueller-Hinton II (MH-II) Agar is a non-selective and non-differential medium. Originally developed by Mueller and Hinton as a transparent medium for isolating Neisseria species, MH-II uses starch in place of pea extract to protect against toxic substances. Bauer, Kirby, Sherris, and Tuck recommended the medium for antibiotic susceptibility testing. The Clinical and Laboratory Standards Institute (CLSI) endorses it because it supports consistent growth of most non-fastidious pathogens, minimizes inhibitors of key antibiotics, and ensures reliable batch-to-batch performance.• Product meets AOAC, ASM, BAM, CLSI, USDA, and WHO performance criteria.• Mueller-Hinton II Agar should be used by trained professionals within a laboratory setting. • Product is not intended for IVD use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.