Learn More
Gawet Marble and Granite Vermont Marble™ Slab, 15″ x 20″ x 2″ thick
Catalog No. NC0078597
Encompass_Preferred
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads
Catalog No. NC0078597 Supplier Gawet Marble and Granite Supplier No. 15X20X2
The Vermont Marble™ Slab is for use on laboratory furniture.   It substantially reduces vibration and shock where a Vermont Marble™ Balance Table cannot be used.– Standard Matte Machined Finish– Includes set of 4 anti-vibration damping pads