Learn More
Chemglass Life Sciences Chemglass Stirrer Bar, Magnetic, PTFE, Double Crosshead, 15 x 22 mm
SDP
Catalog No. NC2057467
Encompass_Preferred
PTFE double crosshead magnetic stirrer bar designed for use in tube-like containers and general laboratory stirring applications. The crosshead shape improves vortex formation and stability while the PTFE coating resists chemical attack and high temperatures.
  • Compact 15 x 22 mm size suitable for small vessels and tubes.
  • PTFE construction provides chemical resistance and temperature stability.
  • Double crosshead shape improves turbulence and mixing efficiency.
  • Magnetic core ensures reliable coupling with magnetic stir plates.
  • Sold individually for flexible replacement and inventory use.
Catalog No. NC2057467 Supplier Chemglass Life Sciences Supplier No. CG2013D04
PTFE double crosshead magnetic stirrer bar designed for use in tube-like containers and general laboratory stirring applications. The crosshead shape improves vortex formation and stability while the PTFE coating resists chemical attack and high temperatures.
  • Compact 15 x 22 mm size suitable for small vessels and tubes.
  • PTFE construction provides chemical resistance and temperature stability.
  • Double crosshead shape improves turbulence and mixing efficiency.
  • Magnetic core ensures reliable coupling with magnetic stir plates.
  • Sold individually for flexible replacement and inventory use.