Learn More
MSC Koch Filter Air Filter, MERV 8, 1 ea
Catalog No. 5000506746
Encompass
This air filter features a wire-backed pleated design, a MERV 8 rating, and media constructed from synthetic material with activated carbon. It has nominal dimensions of 15 in high, 25 in wide, and 1 in deep, and is suitable for a wide range of applications.
  • Wire-backed pleated design for efficient air filtration
  • MERV 8 rating ensures effective particle capture
  • Synthetic media with activated carbon for odor control
  • Beverage board frame provides structural integrity
  • Disposable for convenient replacement
  • Leadership in Energy & Environmental Design certified
  • Constructed with recycled content
  • Withstands maximum temperatures up to 120 °F
  • Complies with ASHRAE 52.2-2012 and UL 900 standards
Catalog No. 50-005-06746 Supplier MSC Supplier No. 76034347
This air filter features a wire-backed pleated design, a MERV 8 rating, and media constructed from synthetic material with activated carbon. It has nominal dimensions of 15 in high, 25 in wide, and 1 in deep, and is suitable for a wide range of applications.
  • Wire-backed pleated design for efficient air filtration
  • MERV 8 rating ensures effective particle capture
  • Synthetic media with activated carbon for odor control
  • Beverage board frame provides structural integrity
  • Disposable for convenient replacement
  • Leadership in Energy & Environmental Design certified
  • Constructed with recycled content
  • Withstands maximum temperatures up to 120 °F
  • Complies with ASHRAE 52.2-2012 and UL 900 standards