Learn More
Ace Glass, Inc. 15mm ID 300mm jacketed chromatography column with #15 Ace-Threds.
SDP
Catalog No. 50972200
Encompass_Preferred
Columns are fully jacketed including internally threaded ends for circulation of heat exchange media. 15mm ID 300mm jacketed chromatography column with #15 Ace- Threds.
Catalog No. 50-972-200 Supplier Ace Glass, Inc. Supplier No. 582113
Columns are fully jacketed including internally threaded ends for circulation of heat exchange media. 15mm ID 300mm jacketed chromatography column with #15 Ace- Threds.