Learn More
Waters Corp Waters Vial, LCGC Certified, Amber Glass, 15 x 45 mm, 4 mL
Catalog No. 50782544 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These certified vials are rigorously tested for cleanliness by HPLC and manufactured to the tightest dimensional tolerances in the industry, ensuring reliable performance for sensitive laboratory applications.
  • LCGC certified for demanding analytical requirements.
  • Made from amber glass for light-sensitive samples.
  • Features a screw neck closure for secure sealing.
  • Dimensions of 15 x 45 mm.
  • Provides a 4 mL volume capacity.
  • Includes a black cap & PTFE/silicone septum.
  • Ideal for sample preparation.
Catalog No. 50-782-544 Supplier Waters Corp Supplier No. 186001133C
These certified vials are rigorously tested for cleanliness by HPLC and manufactured to the tightest dimensional tolerances in the industry, ensuring reliable performance for sensitive laboratory applications.
  • LCGC certified for demanding analytical requirements.
  • Made from amber glass for light-sensitive samples.
  • Features a screw neck closure for secure sealing.
  • Dimensions of 15 x 45 mm.
  • Provides a 4 mL volume capacity.
  • Includes a black cap & PTFE/silicone septum.
  • Ideal for sample preparation.